

Sodium nitrate ≥99.99% (metals basis), Suprapur® for trace analysis, Supelco®
Catalog # 1.06546.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:874 g/l (20 °C)
- Melting Point:308 °C
- Molar Mass:84.99 g/mol
- Bulk Density:1200 kg/m3
- Density:2.26 g/cm3 (20 °C)
- pH:5.5 - 8.0 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:84.99 g/mol
Merck Index:13,08720
Boiling Pt:380 °C (1013 hPa)
Melting Pt:308 °C
Density:2.26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-554-3
UN:1498
ADR:5.1,III
:M-Clarity™ quality level = MQ100
REACH:01-2119488221-41
:GHS03
:GHS07
CAS Number:7631-99-4
Formula:NaNO₃
MDL Number:MFCD00011119