Lithium nitrate ≥99%, purified
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7790-69-4
:
Formula:LiNO₃
MDL Number:MFCD00011094
:68.95 g/mol
Merck Index:13,05558
Melting Pt:255 °C
Density:2.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-218-9
UN:2722
ADR:5.1,III
:GHS03
Specification Test Results
- Assay:Min. 99 %
- Cl (Chloride):Max. 200 ppm
- SO4 (Sulphate):Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Number750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC25029.268 spec_VWRC25029.268 spec_VWRC25029.360
Product specification
Frequently Bought Together