

Lithium nitrate, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-69-4
:
Formula:LiNO₃
MDL Number:MFCD00011094
:68.95 g/mol
Merck Index:14,5536
Boiling Pt:600 °C (dec.)
Melting Pt:264 °C
Density:2.38
Storage Temperature:Ambient
EINECS:232-218-9
UN:2722
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together