Lithium nitrate 99+%, extra pure
:7790-69-4
:
:LiNO₃
:MFCD00011094
:68.95 g/mol
:13,05558
:251 °C
:2.38 g/cm³
:Ambient
:232-218-9
:2722
:5.1,III
:GHS07
:GHS03
- Appearance:White Adhering crystalline powder
- Loss on drying:≤1 % (200°C)
- Titration after Ion exchange:≥99.0 %
Frequently Bought Together