Order Entry
Switzerland
ContactUsLinkComponent
 
 
Potassium nitrate, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
 
Potassium nitrate, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    320 g/l (20 °C)
  • Melting Point:
    334 °C
  • Molar Mass:
    101.10 g/mol
  • Bulk Density:
    800 kg/m3
  • Density:
    2.11 g/cm3 (20 °C)
  • pH:
    5.0 - 7.5 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

 

Frequently Bought Together