


Potassium nitrite, EMSURE® for analysis, Supelco®
Catalog # 1.05067.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:2810 g/l (20 °C)
- Melting Point:440 °C
- Molar Mass:85.11 g/mol
- Bulk Density:980 kg/m3
- Density:1.92 g/cm3 (20 °C)
- pH:7 - 10 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:85.1 g/mol
Merck Index:13,07734
Melting Pt:387 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-832-4
UN:1488
ADR:5.1,II
:M-Clarity™ quality level = MQ300
:GHS03
:GHS06
:GHS09
CAS Number:7758-09-0
Formula:KNO₂
MDL Number:MFCD00011408