
Sodium periodate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7790-28-5
:
Formula:NaIO₄
MDL Number:MFCD00003534
Merck Index:13,08714
Boiling Pt:300 °C (1013 hPa)
Melting Pt:175 °C
Density:3.87 g/cm³ (20 °C)
EINECS:232-197-6
ADR:5.1,II
:213.89 g/mol
UN:1479
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Cl + ClO3 + Br + BrO3 (as Cl):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Mn (Manganese):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC27986.235 spec_VWRC27986.235 spec_VWRC27986.290 spec_VWRC27986.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together