
Sodium iodate, EMSURE® for analysis, Supelco®
: Merck
:7681-55-2
:
:INaO₃
:MFCD00003533
:13,08704
:420…430 °C
:3.56 g/cm³ (20 °C)
:5.1,II
:M-Clarity™ quality level = MQ300
:197.89 g/mol
:Ambient
:231-672-5
:1479
:GHS03
- Solubility:81 g/l (20 °C)
- Molar Mass:197.89 g/mol
- Bulk Density:1150 kg/m3
- Density:4.28 g/cm3 (20 °C)
- pH:5.8 (90 g/l, H2O, 25 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together