Cover glasses
Supplier: MARIENFELD
Jump to:
About this item
Cover slides suitable for fluorescence microscopy.
- Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
5 Options Available Below
Product Details & Documents
Cover slides suitable for fluorescence microscopy.
- Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- L×WNo.MaterialAvailabilityPrice
- 630-2185High precision cover glassesHigh precision cover glasses18×18 mm#1,5Borosilicate glassLog in for availability1 * 2.000 STSFr. 253.00
- 630-2186High precision cover glassesHigh precision cover glasses22×22 mm#1,5Borosilicate glassLog in for availability1 * 2.000 STSFr. 422.00
- 630-2187High precision cover glassesHigh precision cover glasses24×50 mm#1,5Borosilicate glassLog in for availability1 * 1.000 STSFr. 419.00
- MARI0107242High precision cover glassesHigh precision cover glasses24×60 mm#1,5Borosilicate glassLog in for availability1 * 1.000 STSFr. 496.00
- MARI01079990935High precision cover glasses