

Sodium iodide 99.999% (trace metals basis), extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (760 mmHg)
Melting Pt:661 °C
Density:3.66 g/cm³
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White Granular powder
- Total trace metal impurities:≤0.001 %
Specification Test Results
Frequently Bought Together