ContactUsLinkComponent
Sodium iodide, anhydrous 99+%, pure
Irritant
Environment
Sodium iodide, anhydrous 99+%, pure
Certificates
undefined
Irritant
Environment
Sodium iodide, anhydrous 99+%, pure
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Titration Argentometric:
    ≥99.0 % (on dried substance)
  • Loss on drying:
    ≤3 % (1 g, 105°C, 3 h)