Order Entry
Switzerland
ContactUsLinkComponent
Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®
Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    450 °C
  • Molar Mass:
    133.84 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1180 °C
  • Density:
    4 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together