Order Entry
Switzerland
ContactUsLinkComponent
Potassium iodide 99.0-100.5% (by argentometric analysis, on dry substance), EMPROVE® api Ph. Eur., BP, JP, USP, SAFC®
Health
Potassium iodide 99.0-100.5% (by argentometric analysis, on dry substance), EMPROVE® api Ph. Eur., BP, JP, USP, SAFC®
Certificates
Health
Potassium iodide 99.0-100.5% (by argentometric analysis, on dry substance), EMPROVE® api Ph. Eur., BP, JP, USP, SAFC®
Certificates

Specification Test Results

  • Assay (argentometric; calculated on dried substance):
    99.0 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Appearance of solution (JP):
    passes test
  • Alkaline impurities:
    passes test
  • Chloride, Bromide and Thiosulfate:
    passes test
  • Cyanide (CN):
    passes test
  • Iodate(IO₃):
    passes test
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Thiosulfate (S₂O₃):
    passes test
  • Thiosulfate and barium:
    passes test
  • Total nitrogen (N):
    ≤ 0.001 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • As (Arsenic):
    ≤ 0.0003 %
  • Fe (Iron):
    ≤ 0.0002 %
  • Na (Sodium):
    ≤ 0.1 %
  • Residual solvents (Ph Eur/USP/ICH) class 3:
    ≤ 0.1 %
  • Other residual solvents (Ph Eur/USP/ICH):
    excluded by manufacturing process
  • Loss on Drying (105 °C):
    ≤ 0.5 %

Specification Test Results

Related Information

Frequently Bought Together