Order Entry
Switzerland
ContactUsLinkComponent
Sodium fluoride 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur.
Toxic
Sodium fluoride 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Sodium fluoride 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on dried substance):
    99.0 - 100.5 %
  • Appearance of solution:
    Passes test Ph.Eur.
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Identification C:
    Passes test Ph.Eur.
  • Acidity:
    ≤ 0.030 meq/g
  • Alkalinity:
    ≤ 0.010 meq/g
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Insolubility in water:
    ≤ 100 ppm
  • Loss on drying (150°C):
    ≤ 0.2 %
  • Sodium fluosilicate (Na2SiF6):
    ≤ 0.1 %
  • Cl (Chloride):
    ≤ 100 ppm
  • SO3 (Sulphite):
    ≤ 50 ppm
  • SO4 (Sulphate):
    ≤ 100 ppm
  • Fe (Iron):
    ≤ 20 ppm
  • K (Potassium):
    ≤ 0.02 %
  • Conforms to Reag. Ph.Eur:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together