Order Entry
Switzerland
ContactUsLinkComponent
 
 
Sodium fluoride, Suprapur® for trace analysis, Supelco®
  1.06450.0025
 :  Merck
 :  
undefined
 
Sodium fluoride, Suprapur® for trace analysis, Supelco®
  1.06450.0025
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Solubility:
    42 g/l (20 °C)
  • Melting Point:
    993 °C
  • Molar Mass:
    41.98 g/mol
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    1704 °C
  • Vapor Pressure:
    1 hPa (1077 °C)
  • Density:
    2.8 g/cm3 (20 °C)
  • pH:
    10.2 (40 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

 

 

 :
 :41.99 g/mol
 :13,08691
 :1700 °C (1013 hPa)
 :993 °C
 :2.8 g/cm³ (20 °C)
 :Ambient
 :231-667-8
 :1690
 :6.1,III
 :M-Clarity™ quality level = MQ100
 :GHS06
 :7681-49-4
 :NaF
 :MFCD00003524