Order Entry
Switzerland
ContactUsLinkComponent
Potassium chromate in aqueous solution
Health
Irritant
Environment
Potassium chromate in aqueous solution
Catalog # HACH16649H
Supplier:  Hach
CAS Number:  
Health
Irritant
Environment
Potassium chromate in aqueous solution
Catalog # HACH16649H
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :194.19 g/mol
Merck Index:12,07784
Melting Pt:985 °C
Storage Temperature:Ambient
EINECS:232-140-5
UN:3287
ADR:6.1,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:7789-00-6
Formula:K₂CrO₄
MDL Number:MFCD00011368