Overhead stirrer tools, paddle stirrers, BOLA
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with two 45° angled blades.
- Creates axial flow
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Frequently Bought Together