Order Entry
Switzerland
ContactUsLinkComponent
 
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, EMSURE® ACS, ISO for analysis (max. 0.01 ppm Hg), Supelco®
Catalog # 1.04865.0500
Supplier:  Merck
CAS Number:  
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, EMSURE® ACS, ISO for analysis (max. 0.01 ppm Hg), Supelco®
Catalog # 1.04865.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Solubility:
    130 g/l (20 °C)
  • Melting Point:
    398 °C
  • Molar Mass:
    294.19 g/mol
  • Bulk Density:
    1250 kg/m3
  • Boiling Point:
    500 °C (1013 hPa)
  • Density:
    2.69 g/cm3 (20 °C)
  • pH:
    3.6 (100 g/l, H2O)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :294.18 g/mol
Merck Index:12,07790
Boiling Pt:500 °C (1013 hPa)
Melting Pt:398 °C
Density:2.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-906-6
UN:3086
ADR:6.1 (5.1),II
 :M-Clarity™ quality level = MQ300
REACH:01-2119454792-32
 :GHS03
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367