

Lithium perchlorate, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-03-9
:
Formula:ClLiO₄
MDL Number:MFCD00011079
:106.39 g/mol
Merck Index:14,5539
Boiling Pt:400 °C (dec.)
Melting Pt:236 °C
Density:2.43
UN:1481
ADR:5.1,II
:ampouled under argon
EINECS:232-237-2
:GHS03
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together