Order Entry
Switzerland
ContactUsLinkComponent
Lithium perchlorate 95+% ACS
Irritant
Oxidizing
Lithium perchlorate 95+% ACS
Catalog # ACRO423865000
CAS Number:  
undefined
Irritant
Oxidizing
Lithium perchlorate 95+% ACS
Catalog # ACRO423865000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Crystalline or granular powder or crystals
  • Titration Argentometric:
    ≥95.0 %
  • pH:
    6.0 to 7.5 (5% soln. at 25°C)
  • Chloride (Cl):
    ≤0.003 %
  • Sulfate (SO4):
    ≤0.001 %
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (ICP-OES):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

About this item

 :
 :106.39 g/mol
Merck Index:13,05561
Boiling Pt:430 °C (1013 hPa)
Melting Pt:236 °C
Density:2,42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-237-2
UN:1481
ADR:5.1,II
 :GHS07
 :GHS03
CAS Number:7791-03-9
Formula:ClLiO₄
MDL Number:MFCD00011079