Jars, with screw cap and tubing connections, BOLA
Jars, made of PFA, transparent with screw-cap.
- Sturdy design
- Non-porous
- Good chemical resistance
- Temperature resistance from −200 to +250 °C
- FDA conform
Tubings with an external diameter of max. 6,35 mm can be inserted through the screw caps.
Frequently Bought Together