Jars, with screw cap, BOLA
Supplier: Bohlender
Certificates
About this item
Made of PFA, transparent with screw-cap.
- Sturdy design
- Non-porous
- Good chemical resistance
- Temperature resistance from −200 to +250 °C
Frequently Bought Together
Made of PFA, transparent with screw-cap.