Order Entry
Switzerland
ContactUsLinkComponent
Sodium chloride for spectroscopy
Sodium chloride for spectroscopy
Catalog # 43631.19
CAS Number:  
Sodium chloride for spectroscopy
Catalog # 43631.19
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    75 g
  • Seq:
    3389

Specifications

About this item

 :
 :58.44 g/mol
Merck Index:14,8599
Boiling Pt:1413 °C
Melting Pt:801 °C
Density:2.165
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477