Order Entry
Switzerland
ContactUsLinkComponent
Lithium chloride ≥98%, purified
Irritant
Lithium chloride ≥98%, purified
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Lithium chloride ≥98%, purified
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Frequently Bought Together