Potassium chloride, Ultrapure for spectroscopy
Catalog # 39795.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Potassiumchloride
:
:74.55 g/mol
Merck Index:14,7621
Boiling Pt:1500 °C (subl.)
Melting Pt:773 °C
Density:1.984
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360