Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
:144-55-8
:
:NaHCO₃
:MFCD00003528
:84.01 g/mol
:13,08655
:270 °C
:2.159 g/cm³ (20 °C)
:Ambient
:205-633-8
- Assay (calc. on dried substance):99.7 - 101,0 %
- Appearance of solution:Passes test Ph.Eur.
- Carbonates:Passes test Ph.Eur.
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Identification C:Passes test Ph.Eur.
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 0.015 %
- Loss on drying:Max. 0.2 %
- Reducing substances (as I):Max. 65 ppm
- Total N (Nitrogen):Max. 5 ppm
- Total S (as SO4):Max. 30 ppm
- Cl (Chloride):Max. 50 ppm
- NH4 (Ammonium):Max. 5 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- As (Arsenic):Max. 2 ppm
- Ca (Calcium):Max. 100 ppm
- Cu (Copper):Max. 2 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 50 ppm
- Pb (Lead):Max. 5 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 33L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Frequently Bought Together