Sodium hydrogen carbonate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Assay (dried basis):99.7 - 100.3 %
- Calcium (Ca):max. 0.02 %
- Chloride (Cl):max. 0.003 %
- Iron (Fe):max. 0.001 %
- Potassium (K):max. 0.005 %
- Magnesium (Mg):max. 0.005 %
- Insoluble Matter:max. 0.015 %
- Phosphate (PO4):max. 0.001 %
- Sulfur Compounds (as SO4):max. 0.003 %
- Ammonium (NH4):max. 5
- Heavy Metals (as Pb):max. 5
Specification Test Results
Frequently Bought Together