
Sodium carbonate ≥99.9%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Specification Test Results
- Assay (calculated on dried substance):Min. 99.9 %
- Heavy metals (as Pb):Max. 5 ppm
- Insoluble substances:Max. 100 ppm
- Loss on drying (300 °C):Max. 1 %
- Total N (Nitrogen):Max. 10 ppm
- Total S (as SO4):Max. 30 ppm
- Cl (Chloride):Max. 10 ppm
- PO4 (Phosphate):Max. 10 ppm
- SiO4 (as SiO2):Max. 20 ppm
- Al (Aluminium):Max. 10 ppm
- Ca (Calcium):Max. 100 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 50 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
27771_Sodium carbonate anhydrous_BSE_Rev0
BSE/TSE Certificates
spec_VWRC27771.233 spec_VWRC27771.260 spec_VWRC27771.260 spec_VWRC27771.290 spec_VWRC27771.360
Product specification
More
Frequently Bought Together