Orders LinkContactUsLinkComponent
Spoon spatulas
Spoon spatulas
Supplier:   RSG
Spoon spatulas
Supplier:  RSG
Certificates
Spoon spatulas
4 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
  • With round handle
  • Very good thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 150 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 73.70
    SFr. 73.70
    1 * 5 ST
       SFr. 73.70
  • 180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 77.40
    SFr. 77.40
    1 * 5 ST
       SFr. 77.40
  • 210 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 84.20
    SFr. 84.20
    1 * 5 ST
       SFr. 84.20
  • 250 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 88.60
    SFr. 88.60
    1 * 5 ST
       SFr. 88.60