
Lithium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.05680.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:13 g/l (20 °C)
- Melting Point:720 °C
- Molar Mass:73.89 g/mol
- Bulk Density:250 kg/m3
- Vapor Pressure:(20 °C)
- Density:2.1 g/cm3 (20 °C)
- pH:10 - 11 (5 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2.11 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:209-062-5
:GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084