Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium bromide ≥98%, GPR RECTAPUR®
  27742.290
 :  
undefined
Sodium bromide ≥98%, GPR RECTAPUR®
  27742.290
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay (calculated on dried substance):
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Loss on drying (150°C):
    Max. 2 %
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 50 ppm

 

 

 :
 :102.89 g/mol
 :13,08666
 :1396 °C (1013 hPa)
 :753 °C
 :3.2 g/cm³ (20 °C)
 :Ambient
 :231-599-9
 :01-2119490106-41
 :7647-15-6
 :NaBr
 :MFCD00003475