Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium bromide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
 
Sodium bromide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
 

 

  • Assay (calculated to the dried substance):
    98.5 - 100.5 %
  • Loss on drying (105°C, 3h):
    Max. 1.0 %
  • Acidity or alkalinity:
    complying
  • Arsenic (As):
    Max. 0.0002 %
  • Barium (Ba):
    Max. 0.002 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Bromate (BrO₃):
    complying
  • Chloride (Cl):
    Max. 0.2 %
  • Iodide (I):
    complying
  • Sulfate (SO₄):
    Max. 0.005 %
  • Mg, earth alkali metals (as Ca):
    Max. 200 ppm
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

 

 

Frequently Bought Together