
Lithium bromide ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077
:86.84 g/mol
Merck Index:13,05548
Boiling Pt:1.265 °C (1.013 hPa)
Melting Pt:550 °C
Density:3.46 g/cm3 (20 °C)
EINECS:231-439-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay of Br:91.1 - 92.9 %
- Appearance of the solution:complying
Specification Test Results
Frequently Bought Together