Order Entry
Switzerland
ContactUsLinkComponent
 
 
Lithium bromide ≥99%, GPR RECTAPUR®
  25002.266
 :  
undefined
 
Lithium bromide ≥99%, GPR RECTAPUR®
  25002.266
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 99 %
  • SO4 (Sulphate):
    Max. 0.04 %
  • Fe (Iron):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 0.1 %
  • Ba (Barium):
    Max. 0.02 %
  • Ca (Calcium):
    Max. 100 ppm
  • K (Potassium):
    Max. 0.02 %
  • Mg (Magnesium):
    Max. 10 ppm
  • Na (Sodium):
    Max. 0.2 %
  • Pb (Lead):
    Max. 10 ppm

 

 

 :
 :86.84 g/mol
 :13,05548
 :1265 °C (1013 hPa)
 :550 °C
 :3.46 g/cm³ (20 °C)
 :Ambient
 :231-439-8
 :GHS07
 :7550-35-8
 :23303-71-1
 :LiBr
 :MFCD00011077