


Lithiumtetrahydroborat 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16949-15-8
:
Formula:H₄BLi
MDL Number:MFCD00011088
:21.78 g/mol
Merck Index:13,05547
Boiling Pt:66 °C (1013 hPa)
Melting Pt:280 °C
Density:0.66 g/cm³
Storage Temperature:Raumtemperatur
EINECS:241-021-7
UN:1413
ADR:4.3,I
:GHS06
:GHS05
:GHS02
Specification Test Results
- Appearance:White Powder
- Assay:≥94 %
Specification Test Results
Frequently Bought Together