di-Sodium tetraborate, anhydrous ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1330-43-4
:
Formula:Na₂B₄O₇
MDL Number:MFCD00081185
:201.22 g/mol
Merck Index:14,8590
Boiling Pt:1575 °C (dec.)
Melting Pt:741 °C
Density:2.367
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
Frequently Bought Together