Order Entry
Switzerland
ContactUsLinkComponent
di-Lithium tetraborate for analysis, Supelco®
Corrosive
Health
Irritant
di-Lithium tetraborate for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Health
Irritant
di-Lithium tetraborate for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Assay (acidimetric):
    ≥ 98.0 %
  • Chloride (Cl):
    ≤ 0.005 %
  • Phosphate (PO₄):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Heavy metals (as Pb):
    ≤ 0.005 %
  • Ca (Calcium):
    ≤ 0.005 %
  • Fe (Iron):
    ≤ 0.001 %
  • K (Potassium):
    ≤ 0.005 %
  • Mg (Magnesium):
    ≤ 0.001 %
  • Na (Sodium):
    ≤ 0.005 %

Specification Test Results

Related Information

Frequently Bought Together