


1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1165
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
EINECS:204-661-8
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay:Min. 99 %
- Boiling point:99 - 102 °C
- Density (20/4):1.030 - 1.035
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
Frequently Bought Together