


1,4-Dioxane stabilised, EMSURE® ACS, ISO for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,
:M-Clarity™ quality level = MQ300
Flash Pt:12 °C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
EINECS:204-661-8
UN:1165
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:12 °C
- Molar Mass:88.11 g/mol
- Boiling Point:101.5 °C (1013 hPa)
- Vapor Pressure:41 hPa (20 °C)
- Flash Point:11 °C
- Explosion Limit:1.7 - 25.2 %(V)
- Density:1.03 g/cm3 (20 °C)
- pH:6 - 8 (500 g/l, H2O, 20 °C)
- Ignition Point:300 °C
- Evaporation number:7.3
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.09671.2500_TDS_EN ProductPage_109671
Product specification
msds_109671
SDS
Frequently Bought Together