Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
1,4-Dioxane ≥99.5% (by GC) stabilised, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
 
undefined
 
 
 
1,4-Dioxane ≥99.5% (by GC) stabilised, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99.5 %
  • Congealing - Freezing Point:
    Min. 11 °C
  • Boiling range:
    100 - 102 °C
  • Density (D 20/4):
    1.031 - 1.034
  • Refractive index (n 20/D):
    1.4210 - 1.4240
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Acetaldehyde:
    Max. 0.005 %
  • Carbonyl Compounds (as HCHO):
    Max. 0.01 %
  • Peroxides (as H₂O₂):
    Max. 0.005 %
  • APHA:
    Max. 10

 

 

Frequently Bought Together