Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
1,4-Dioxane
1,4-Dioxane 99+% ACS
1,4-Dioxane 99+% ACS
:
Thermo Fisher Scientific
1,4-Dioxane 99+% ACS
:
Thermo Fisher Scientific
:
123-91-1
:
:
C₄H₈O₂
:
MFCD00006571
:
88.11 g/mol
:
14,3300
:
100…102 °C
:
11.8 °C
:
1.034
:
12 °C (54 °F)
:
Ambient
:
204-661-8
:
1165
:
3,II
:
GHS02
:
GHS08
:
GHS07
msds_ALFA039118
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
: Merck
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Petroleum spirit, 100…120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media
: Thermo Fisher Scientific