Order Entry
Switzerland
ContactUsLinkComponent
Cyclopentane for synthesis, Sigma-Aldrich®
Cyclopentane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Cyclopentane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -93.3 °C
  • Molar Mass:
    70.13 g/mol
  • Boiling Point:
    49 °C
  • Vapor Pressure:
    346 hPa (20 °C)
  • Flash Point:
    -42 °C
  • Refractive Index:
    1.4065 (20 °C)
  • Explosion Limit:
    1.5 - 8.7 %(V)
  • Density:
    0.75 g/cm3 (20 °C)
  • Ignition Point:
    380 °C

Specification Test Results

Related Information

Frequently Bought Together