Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexane, Supelco® | Avantor

Cyclohexane, Supelco®

Flammable
Health
Irritant
Environment
Supplier:   Merck
Cyclohexane, Supelco®
Supplier:  Merck
Certificates
Cyclohexane, Supelco®
Cyclohexane, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-806-2

Dokumente

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - SFr. 98.10
    SFr. 98.10
    1 * 5 mL
       SFr. 98.10

  • Glass bottle
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - SFr. 164.00
    SFr. 164.00
    1 * 10 mL
       SFr. 164.00