Supelco® Organic Reference Standard, Acetoin
Catalog # 40127-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:513-86-0
- Description:Acetoin
- Storage conditions:2...8 °C
- Hazard class description:H228 - H315 - H319
- Shipping conditions:STD
- MDL No.:MFCD00004521
- M-Clarity quality level:MQ100
- Synonym(s):3-Hydroxy-2-butanone; Acetylmethylcarbinol
- Melting point:15 °C (monomer); 90 °C (dimer) (lit.)
- Formula:CH₃COCH(OH)CH₃
- Molecular weight:88,11
- Density:1,013 g/ml at 25 °C (lit.)
- Flash point:47 °C
- Boiling point:148 °C (lit.)
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P240 - P241 - P264 - P302 + P352 - P305 + P351 + P338