
2-Chloroethyldiisopropylammonium chloride, Sigma-Aldrich®
: Merck
:4261-68-1
:
:C₈H₁₉Cl₂N
:MFCD00012496
:200.15 g/mol
:6.1,I
:M-Clarity™ quality level = MQ200
:224-238-1
:2811
:GHS06
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:130 - 133 °C
- Molar Mass:200.15 g/mol
- pH:4.0 - 4.5 (10 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together