Saccharin sodium dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6155-57-3
:
Formula:C₇H₅NNaO₃S·2H₂O
MDL Number:MFCD00151213
:242.21 g/mol
:M-Clarity™ quality level = MQ200
EINECS:204-886-1
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Molar Mass:241.19 g/mol
- Bulk Density:600 - 800 kg/m3
- Flash Point:200 °C
- pH:6.0 - 7.5 (100 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together