Organic reference standard, Bis(2-ethylhexyl) phthalate 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL16173000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:117-81-7
- Description:Bis(2-ethylhexyl) phthalate 10 µg/ml in cyclohexane
- Formula:C₂₄H₃₈O₄
- Description:Bis(2-ethylhexyl) phthalate