Cyclohexane for pesticide residue analysis
:110-82-7
:
:C₆H₁₂
:MFCD00003814
:84.16 g/mol
:13,02752
:81 °C (1013 hPa)
:6.5 °C
:0.77 g/cm³
:–18 °C
:203-806-2
:1145
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- GC:(ECD) Passes test, (NPD) Passes test, (FID) Passes test
- Residue after evaporation:≤2 ppm
Frequently Bought Together