



Cyclohexan 99.5% zur Analyse
:110-82-7
:
:C₆H₁₂
:MFCD00003814
:84.16 g/mol
:13,02752
:81 °C (1013 hPa)
:6.5 °C
:0.77 g/cm³
:–18 °C
:203-806-2
:1145
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Raumtemperatur
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Potassium (K):≤0.2 ppm
- Sodium (Na):≤1 ppm
- Zinc (Zn):≤0.1 ppm
Frequently Bought Together