



Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:6.5 °C
Density:0.77 g/cm³
Flash Pt:–18 °C
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 %
- GC:≥99.0 %
- UV:at 210 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0002 meq/g
Specification Test Results
Frequently Bought Together